Product Details

SNP ID
rs147776879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43726376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACATGCTGGCAATTAGCGCCAT[C/T]GACATTCCAGATATCGGGGCTGGGA
Phenotype
MIM: 601415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTSF1L PubMed Links

Gene Details

Gene
GTSF1L
Gene Name
gametocyte specific factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008901.1 627 Intron NP_001008901.1
NM_176791.3 627 Missense Mutation AAT,GAT N107D NP_789761.1
XM_005260298.4 627 Intron XP_005260355.1
Gene
LOC101927200
Gene Name
uncharacterized LOC101927200
There are no transcripts associated with this gene.

Gene
MYBL2
Gene Name
MYB proto-oncogene like 2
There are no transcripts associated with this gene.

View Full Product Details