Product Details
- SNP ID
-
rs150317541
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35627306 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCAGTCCCCTGATGTGCAGCCT[C/G]GGCTGCAAACCTGGCCACATGGTTG
- Phenotype
-
MIM: 604205
MIM: 603038
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147019139] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPNE1
- Gene Name
- copine 1
- Gene
- SPAG4
- Gene Name
- sperm associated antigen 4
There are no transcripts associated with this gene.
View Full Product Details