Product Details

SNP ID
rs151053015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62410929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGAGGGACTGTGGTCCTCGGC[A/G]TCCCTTGGGCTCCCGGGCCCCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABLES2 PubMed Links

Gene Details

Gene
CABLES2
Gene Name
Cdk5 and Abl enzyme substrate 2
There are no transcripts associated with this gene.

Gene
RBBP8NL
Gene Name
RBBP8 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080833.2 2141 Silent Mutation GAC,GAT D648D NP_543023.2

View Full Product Details