Product Details

SNP ID
rs149397253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44504807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATAATACTGTTCACCGAGAAG[C/G]TACGCCCATCGAAGGAGTTGGCCAC
Phenotype
MIM: 612920
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSPEAR PubMed Links

Gene Details

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 1957 Missense Mutation ACC,AGC T542S NP_001258966.1
NM_144991.2 1957 Missense Mutation ACC,AGC T610S NP_659428.2
Gene
TSPEAR-AS1
Gene Name
TSPEAR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details