Product Details

SNP ID
rs149543089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14485908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGATATATAGTTGGGAATGC[A/G]TGTTCAGTCACTTGGCTCTGTGATA
Phenotype
MIM: 607978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMSN1 PubMed Links

Gene Details

Gene
SAMSN1
Gene Name
SAM domain, SH3 domain and nuclear localization signals 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256370.1 1265 UTR 3 NP_001243299.1
NM_001286523.1 1265 UTR 3 NP_001273452.1
NM_022136.4 1265 UTR 3 NP_071419.3
XM_011529684.2 1265 UTR 3 XP_011527986.1
XM_011529685.1 1265 UTR 3 XP_011527987.1
XM_011529686.1 1265 Intron XP_011527988.1
XM_017028427.1 1265 Intron XP_016883916.1

View Full Product Details