Product Details

SNP ID
rs140565519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23764861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGGAAGCGTATGGGCACTCGG[C/T]GAGGCCGCCATGAGCAAAGCCCCAC
Phenotype
MIM: 615903 MIM: 185261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182520.2 1182 Nonsense Mutation CGA,TGA R132* NP_872326.2
XM_011529907.2 1182 Nonsense Mutation CGA,TGA R202* XP_011528209.1
XM_011529908.2 1182 Nonsense Mutation CGA,TGA R137* XP_011528210.2
XM_011529912.2 1182 Nonsense Mutation CGA,TGA R149* XP_011528214.1
XM_017028602.1 1182 Nonsense Mutation CGA,TGA R230* XP_016884091.1
XM_017028603.1 1182 Nonsense Mutation CGA,TGA R223* XP_016884092.1
XM_017028604.1 1182 Nonsense Mutation CGA,TGA R210* XP_016884093.1
XM_017028605.1 1182 Nonsense Mutation CGA,TGA R207* XP_016884094.1
XM_017028606.1 1182 Nonsense Mutation CGA,TGA R230* XP_016884095.1
XM_017028607.1 1182 Nonsense Mutation CGA,TGA R230* XP_016884096.1
XM_017028608.1 1182 Nonsense Mutation CGA,TGA R230* XP_016884097.1
XM_017028609.1 1182 Nonsense Mutation CGA,TGA R167* XP_016884098.1
XM_017028610.1 1182 Nonsense Mutation CGA,TGA R160* XP_016884099.1
XM_017028611.1 1182 Silent Mutation GGC,GGT G203G XP_016884100.1
XM_017028612.1 1182 Nonsense Mutation CGA,TGA R132* XP_016884101.1
XM_017028613.1 1182 Intron XP_016884102.1
XM_017028614.1 1182 Intron XP_016884103.1
XM_017028615.1 1182 Silent Mutation GGC,GGT G133G XP_016884104.1
XM_017028616.1 1182 Intron XP_016884105.1
XM_017028617.1 1182 Intron XP_016884106.1
XM_017028618.1 1182 Intron XP_016884107.1
Gene
CHCHD10
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301339.1 1182 Intron NP_001288268.1
NM_213720.2 1182 Intron NP_998885.1
Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

View Full Product Details