Product Details

SNP ID
rs147366066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38216440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATTTCCTGACTTACAGCAAGC[A/G]AGTTATCGTCTTCTGTATTTTGTAG
Phenotype
MIM: 604877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFF PubMed Links

Gene Details

Gene
MAFF
Gene Name
MAF bZIP transcription factor F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161572.1 1445 UTR 3 NP_001155044.1
NM_001161573.1 1445 UTR 3 NP_001155045.1
NM_001161574.1 1445 UTR 3 NP_001155046.1
NM_012323.3 1445 UTR 3 NP_036455.1
Gene
SNORA92
Gene Name
small nucleolar RNA, H/ACA box 92
There are no transcripts associated with this gene.

Gene
TMEM184B
Gene Name
transmembrane protein 184B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195071.1 1445 Intron NP_001182000.1
NM_001195072.1 1445 Intron NP_001182001.1
NM_012264.4 1445 Intron NP_036396.2
XM_011530112.2 1445 Intron XP_011528414.2
XM_011530113.1 1445 Intron XP_011528415.1
XM_011530114.1 1445 Intron XP_011528416.1
XM_011530115.2 1445 Intron XP_011528417.1
XM_011530116.2 1445 UTR 3 XP_011528418.2
XM_011530117.1 1445 Intron XP_011528419.1
XM_011530118.1 1445 Intron XP_011528420.1
XM_011530119.1 1445 Intron XP_011528421.1
XM_017028754.1 1445 Intron XP_016884243.1
XM_017028755.1 1445 Intron XP_016884244.1
XM_017028756.1 1445 Intron XP_016884245.1
XM_017028757.1 1445 Intron XP_016884246.1
XM_017028758.1 1445 UTR 3 XP_016884247.1
XM_017028759.1 1445 Intron XP_016884248.1

View Full Product Details