Product Details
- SNP ID
-
rs148359937
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:24183531 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATCCAGACCCTCGGCCATGTGGA[C/T]TCCTCCGGGCAAGTGCACTGTGTGT
- Phenotype
-
MIM: 604251
MIM: 615825
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CABIN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9680526] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CABIN1
- Gene Name
- calcineurin binding protein 1
There are no transcripts associated with this gene.
- Gene
- SUSD2
- Gene Name
- sushi domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019601.3 |
368 |
Silent Mutation |
GAC,GAT |
D108D |
NP_062547.1 |
View Full Product Details