Product Details

SNP ID
rs148467697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24184156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGCACCCCAACAAAGTGTCA[A/G]TGATGGAGAAGAGCGAGTTGGTGAA
Phenotype
MIM: 604251 MIM: 615825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABIN1 PubMed Links

Gene Details

Gene
CABIN1
Gene Name
calcineurin binding protein 1
There are no transcripts associated with this gene.

Gene
SUSD2
Gene Name
sushi domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019601.3 504 Missense Mutation ATG,GTG M154V NP_062547.1

View Full Product Details