Product Details

SNP ID
rs138039736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:94871641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTACGACGTGGCCCGTAACA[C/T]GGGCGCCTACACGTCCTCCGGCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC442028 PubMed Links

Gene Details

Gene
LOC442028
Gene Name
uncharacterized LOC442028
There are no transcripts associated with this gene.

Gene
TEKT4
Gene Name
tektin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286559.1 Intron NP_001273488.1
NM_144705.3 Intron NP_653306.1
XM_005263876.2 Intron XP_005263933.1
XM_011510670.1 Intron XP_011508972.1

View Full Product Details