Product Details

SNP ID
rs140813639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201185839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAAAACTGTAAAGTGAGCTTTC[A/G]TGAGAAGCTTCTGATTATTGATTCA
Phenotype
MIM: 601762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP10 PubMed Links

Gene Details

Gene
CASP10
Gene Name
caspase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206524.1 480 Missense Mutation CAT,CGT H21R NP_001193453.1
NM_001206542.1 480 Missense Mutation CAT,CGT H21R NP_001193471.1
NM_001230.4 480 Missense Mutation CAT,CGT H21R NP_001221.2
NM_001306083.1 480 Missense Mutation CAT,CGT H21R NP_001293012.1
NM_032974.4 480 Missense Mutation CAT,CGT H21R NP_116756.2
NM_032976.3 480 Missense Mutation CAT,CGT H21R NP_116758.1
NM_032977.3 480 Missense Mutation CAT,CGT H21R NP_116759.2
XM_005246907.2 480 Missense Mutation CAT,CGT H21R XP_005246964.1
XM_006712796.3 480 UTR 5 XP_006712859.1
XM_011511990.2 480 Missense Mutation CAT,CGT H21R XP_011510292.1

View Full Product Details