Product Details

SNP ID
rs141841093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97124550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTGATGTGCTTTCTCGAGATGC[A/G]TTTCGAAAGATTGCAGGAGATTATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD36 PubMed Links

Gene Details

Gene
ANKRD36
Gene Name
ankyrin repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164315.1 928 Silent Mutation NP_001157787.1
XM_006712514.3 928 Intron XP_006712577.1
XM_011511130.1 928 Intron XP_011509432.1
XM_011511131.1 928 Intron XP_011509433.1
XM_017004009.1 928 Intron XP_016859498.1
XM_017004010.1 928 Intron XP_016859499.1
XM_017004011.1 928 Intron XP_016859500.1
XM_017004012.1 928 Intron XP_016859501.1
XM_017004013.1 928 Intron XP_016859502.1
XM_017004014.1 928 Intron XP_016859503.1
XM_017004015.1 928 Intron XP_016859504.1
XM_017004016.1 928 Intron XP_016859505.1
XM_017004017.1 928 Intron XP_016859506.1
XM_017004018.1 928 Intron XP_016859507.1
XM_017004019.1 928 Intron XP_016859508.1
XM_017004020.1 928 Intron XP_016859509.1
XM_017004021.1 928 Intron XP_016859510.1
XM_017004022.1 928 Intron XP_016859511.1
XM_017004023.1 928 Intron XP_016859512.1
XM_017004024.1 928 Intron XP_016859513.1
XM_017004025.1 928 Intron XP_016859514.1
XM_017004026.1 928 Intron XP_016859515.1
XM_017004027.1 928 Intron XP_016859516.1
XM_017004028.1 928 Intron XP_016859517.1
XM_017004029.1 928 Intron XP_016859518.1
XM_017004030.1 928 Intron XP_016859519.1
XM_017004031.1 928 Intron XP_016859520.1
XM_017004032.1 928 Intron XP_016859521.1
XM_017004033.1 928 Intron XP_016859522.1
XM_017004034.1 928 Intron XP_016859523.1
XM_017004035.1 928 Intron XP_016859524.1
XM_017004036.1 928 Intron XP_016859525.1
XM_017004037.1 928 Intron XP_016859526.1
XM_017004038.1 928 Intron XP_016859527.1
XM_017004039.1 928 Intron XP_016859528.1
XM_017004040.1 928 Intron XP_016859529.1
XM_017004041.1 928 Intron XP_016859530.1
XM_017004042.1 928 Intron XP_016859531.1
XM_017004043.1 928 Intron XP_016859532.1
XM_017004044.1 928 Intron XP_016859533.1
XM_017004045.1 928 Intron XP_016859534.1
XM_017004046.1 928 Intron XP_016859535.1
XM_017004047.1 928 Intron XP_016859536.1
XM_017004048.1 928 Intron XP_016859537.1
XM_017004049.1 928 Intron XP_016859538.1
XM_017004050.1 928 Intron XP_016859539.1
XM_017004051.1 928 Intron XP_016859540.1

View Full Product Details