Product Details

SNP ID
rs143319872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203439476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTTGGTGTTCTGGTCTCTTTT[G/T]GGGGGAGCAGGAGGGGGTCCTCTCC
Phenotype
MIM: 606442 MIM: 609035
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABI2 PubMed Links

Gene Details

Gene
ABI2
Gene Name
abl-interactor 2
There are no transcripts associated with this gene.

Gene
RAPH1
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203365.3 3795 Intron NP_976241.1
NM_213589.2 3795 Silent Mutation CCG,CCT P1238P NP_998754.1
XM_005246777.3 3795 Silent Mutation CCG,CCT P1290P XP_005246834.1
XM_005246779.4 3795 Silent Mutation CCG,CCT P1290P XP_005246836.1
XM_006712695.3 3795 Silent Mutation CCG,CCT P1290P XP_006712758.1
XM_011511646.2 3795 Silent Mutation CCG,CCT P1290P XP_011509948.1
XM_011511647.1 3795 Silent Mutation CCG,CCT P1265P XP_011509949.1
XM_011511648.1 3795 Silent Mutation CCG,CCT P1263P XP_011509950.1
XM_011511649.2 3795 Intron XP_011509951.1

View Full Product Details