Product Details

SNP ID
rs146789470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31234704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTGAGTGAGGGGCTCAAGAAAC[A/T]CTACAAGAGCAAGCTGCTGCCCTTG
Phenotype
MIM: 610229 MIM: 605891
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145122.1 691 Intron NP_001138594.1
NM_001321270.1 691 Intron NP_001308199.1
XM_011532863.2 691 Intron XP_011531165.1
XM_011532864.2 691 Intron XP_011531166.1
XM_011532865.1 691 Intron XP_011531167.1
XM_011532866.2 691 Intron XP_011531168.1
Gene
EHD3
Gene Name
EH domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014600.2 691 Missense Mutation CAC,CTC H28L NP_055415.1
XM_011532806.2 691 Intron XP_011531108.1

View Full Product Details