Product Details

SNP ID
rs147847809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75493544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGATCCTTATCACCAGAGCAG[C/G]CAGGGTCAGCACCAGCCCGATGCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EVA1A PubMed Links

Gene Details

Gene
EVA1A
Gene Name
eva-1 homolog A, regulator of programmed cell death
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135032.1 629 Missense Mutation CCT,GCT P51A NP_001128504.1
NM_032181.2 629 Missense Mutation CCT,GCT P51A NP_115557.1

View Full Product Details