Product Details

SNP ID
rs150035696
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75493511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCCCGGGACGCCGCCTGCAGT[C/T]TGTGTGGCAAGAGATCCTTATCACC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EVA1A PubMed Links

Gene Details

Gene
EVA1A
Gene Name
eva-1 homolog A, regulator of programmed cell death
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135032.1 662 Missense Mutation AAC,GAC N62D NP_001128504.1
NM_032181.2 662 Missense Mutation AAC,GAC N62D NP_115557.1

View Full Product Details