Product Details

SNP ID
rs138173696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183122827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTAAGGTGAGCGTATGCTGGTA[C/T]GAGGTGTGAGCCTTCAGGCAGAGTG
Phenotype
MIM: 605883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMP3 PubMed Links

Gene Details

Gene
LAMP3
Gene Name
lysosomal associated membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014398.3 2729 UTR 3 NP_055213.2
XM_005247360.4 2729 Intron XP_005247417.1
XM_006713586.3 2729 Intron XP_006713649.1
XM_011512688.1 2729 Intron XP_011510990.1

View Full Product Details