Product Details

SNP ID
rs141184565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49099169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACTCCAGCACAGCCATGGCTC[A/G]TGGGGCTGTGTCATTCAGCACATCA
Phenotype
MIM: 603727 MIM: 614471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6890 PubMed Links

Gene Details

Gene
MIR6890
Gene Name
microRNA 6890
There are no transcripts associated with this gene.

Gene
QARS
Gene Name
glutaminyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272073.1 1775 Missense Mutation NP_001259002.1
NM_005051.2 1775 Missense Mutation NP_005042.1
XM_017006965.1 1775 Nonsense Mutation XP_016862454.1
Gene
QRICH1
Gene Name
glutamine rich 1
There are no transcripts associated with this gene.

Gene
USP19
Gene Name
ubiquitin specific peptidase 19
There are no transcripts associated with this gene.

View Full Product Details