Product Details

SNP ID
rs145300083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184242830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTACCTGAGCAGGTGTGTGAG[A/C]CAGCGTGCAGGCATGGCCCACAGGA
Phenotype
MIM: 608750 MIM: 610145 MIM: 611620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006941.2 1053 Missense Mutation TGG,TGT W331C NP_001006942.1
NM_005787.5 1053 Missense Mutation TGG,TGT W379C NP_005778.1
XM_011512322.1 1053 Missense Mutation TGG,TGT W346C XP_011510624.1
XM_011512323.2 1053 Missense Mutation TGG,TGT W339C XP_011510625.1
Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320373.1 1053 Intron NP_001307302.1
NM_138345.2 1053 Intron NP_612354.1
XM_011513288.2 1053 Intron XP_011511590.1
XM_011513289.2 1053 Intron XP_011511591.1
XM_011513290.2 1053 Intron XP_011511592.1
XM_011513291.2 1053 Intron XP_011511593.1
XM_011513292.2 1053 Intron XP_011511594.1

View Full Product Details