Product Details

SNP ID
rs145589776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151213497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAGTGAATTCTTTCATATACC[G/T]CAAGATTTCTTTTGACTGGCAGCTG
Phenotype
MIM: 611318 MIM: 610116
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR171 PubMed Links

Gene Details

Gene
GPR171
Gene Name
G protein-coupled receptor 171
There are no transcripts associated with this gene.

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1108 Intron NP_443728.3
XM_006713487.3 1108 Intron XP_006713550.1
XM_011512390.2 1108 Intron XP_011510692.1
XM_011512394.2 1108 Intron XP_011510696.1
XM_011512399.2 1108 Intron XP_011510701.1
XM_017005676.1 1108 Intron XP_016861165.1
XM_017005677.1 1108 Intron XP_016861166.1
XM_017005678.1 1108 Intron XP_016861167.1
XM_017005679.1 1108 Intron XP_016861168.1
XM_017005680.1 1108 Intron XP_016861169.1
XM_017005681.1 1108 Intron XP_016861170.1
Gene
P2RY14
Gene Name
purinergic receptor P2Y14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081455.1 1108 Silent Mutation GGG,TGG G274W NP_001074924.1
NM_014879.3 1108 Silent Mutation GGG,TGG G274W NP_055694.3
XM_005247922.3 1108 Missense Mutation GGG,TGG G274W XP_005247979.1
XM_005247923.3 1108 Missense Mutation GGG,TGG G274W XP_005247980.1
XM_011513340.2 1108 Missense Mutation GGG,TGG G274W XP_011511642.1
XM_017007583.1 1108 Missense Mutation GGG,TGG G274W XP_016863072.1

View Full Product Details