Product Details

SNP ID
rs145867435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97943302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAAACACAGAAATGGGACAT[G/T]GAAATATTGTGAGAGAATCAACATC
Phenotype
MIM: 612049
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYBG3 PubMed Links

Gene Details

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153605.3 9068 Missense Mutation ATG,ATT M2967I NP_705833.3
XM_005247117.4 9068 Missense Mutation ATG,ATT M2676I XP_005247174.1
Gene
MINA
Gene Name
MYC induced nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042533.2 9068 UTR 3 NP_001035998.1
NM_001261829.1 9068 UTR 3 NP_001248758.1
NM_032778.5 9068 UTR 3 NP_116167.3
NM_153182.3 9068 UTR 3 NP_694822.2
XM_005247838.4 9068 UTR 3 XP_005247895.1
XM_011513244.2 9068 Intron XP_011511546.1

View Full Product Details