Product Details

SNP ID
rs146191659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45828570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAAACTCACTCTTTAAGGCAG[C/T]GACAGTGTTTTCTAAGTTACTTAAG
Phenotype
MIM: 606568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LZTFL1 PubMed Links

Gene Details

Gene
LZTFL1
Gene Name
leucine zipper transcription factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276378.1 737 Missense Mutation ACT,GCT T199A NP_001263307.1
NM_001276379.1 737 Missense Mutation ACT,GCT T212A NP_001263308.1
NM_020347.3 737 Missense Mutation ACT,GCT T216A NP_065080.1
XM_006713207.2 737 Missense Mutation ACT,GCT T216A XP_006713270.2
XM_011533838.2 737 Missense Mutation ACT,GCT T199A XP_011532140.1
XM_017006644.1 737 Missense Mutation ACT,GCT T199A XP_016862133.1
XM_017006645.1 737 Missense Mutation ACT,GCT T225A XP_016862134.1

View Full Product Details