Product Details

SNP ID
rs146875144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196472138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTCATCTGGGGATCCTTTTTGC[A/C]GGTTTGGCACCATTTGCTCTTTATC
Phenotype
MIM: 612688
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF168 PubMed Links

Gene Details

Gene
RNF168
Gene Name
ring finger protein 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152617.3 1992 Missense Mutation NP_689830.2

View Full Product Details