Product Details

SNP ID
rs148336193
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184054829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGGCCTTCCGTCCATTCACCA[A/G]CTACAGCATCCCTACCCGTGTCAAC
Phenotype
MIM: 610121
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HTR3C PubMed Links

Gene Details

Gene
HTR3C
Gene Name
5-hydroxytryptamine receptor 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130770.2 210 Missense Mutation AAC,AGC N59S NP_570126.2

View Full Product Details