Product Details

SNP ID
rs149112513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97947390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCTTGATCCGGCAGTACTGTGA[A/G]GACAATGTGGTCTTTAAACTGCAGT
Phenotype
MIM: 612049
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBG3 PubMed Links

Gene Details

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
There are no transcripts associated with this gene.

Gene
MINA
Gene Name
MYC induced nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042533.2 1230 Missense Mutation CTC,TTC L374F NP_001035998.1
NM_001261829.1 1230 Missense Mutation CTC,TTC L373F NP_001248758.1
NM_032778.5 1230 Missense Mutation CTC,TTC L373F NP_116167.3
NM_153182.3 1230 Missense Mutation CTC,TTC L374F NP_694822.2
XM_005247838.4 1230 Missense Mutation CTC,TTC L374F XP_005247895.1
XM_011513244.2 1230 Missense Mutation CTC,TTC L143F XP_011511546.1

View Full Product Details