Product Details

SNP ID
rs149224531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120602224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTACAGGATAGGAAAGAAAAAC[G/T]TATCCAGGAAGGAAAATTGGATCGA
Phenotype
MIM: 603840
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDUFB4 PubMed Links

Gene Details

Gene
NDUFB4
Gene Name
NADH:ubiquinone oxidoreductase subunit B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168331.1 1373 UTR 3 NP_001161803.1
NM_004547.5 1373 Missense Mutation CGT,CTT R115L NP_004538.2

View Full Product Details