Product Details

SNP ID
rs149703643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101651568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCGAGCATCTATGTAGGCTTC[A/G]CTACATACAGGACAGACATAGGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB11 PubMed Links

Gene Details

Gene
ZBTB11
Gene Name
zinc finger and BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014415.3 4375 Silent Mutation AGC,AGT S920S NP_055230.2
XM_011512689.2 4375 Silent Mutation AGC,AGT S855S XP_011510991.1
XM_011512690.2 4375 Silent Mutation AGC,AGT S636S XP_011510992.1

View Full Product Details