Product Details
- SNP ID
-
rs138811239
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:24533044 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGTTGTCATAACACCACTGAAC[C/T]GATTCATGATCTAAAAAGGTAGAAG
- Phenotype
-
MIM: 603403
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DHX15
PubMed Links
Gene Details
- Gene
- DHX15
- Gene Name
- DEAH-box helicase 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001358.2 |
2088 |
Silent Mutation |
TCA,TCG |
S640S |
NP_001349.2 |
View Full Product Details