Product Details

SNP ID
rs139062730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67914621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATAAATTCCTTCCATATATCC[A/G]GCACAGAACATTCCAGGTTTTATAT
Phenotype
MIM: 611704
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11A PubMed Links

Gene Details

Gene
TMPRSS11A
Gene Name
transmembrane protease, serine 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114387.1 1183 Silent Mutation GCC,GCT A354A NP_001107859.1
NM_182606.3 1183 Silent Mutation GCC,GCT A357A NP_872412.3

View Full Product Details