Product Details

SNP ID
rs142473295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128884479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGTGAACTATGAATATTTTT[A/G]AATATTTTCCAGATTCATCAGGGAG
Phenotype
MIM: 610514 MIM: 611399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JADE1 PubMed Links

Gene Details

Gene
JADE1
Gene Name
jade family PHD finger 1
There are no transcripts associated with this gene.

Gene
SCLT1
Gene Name
sodium channel and clathrin linker 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300897.1 Intron NP_001287826.1
NM_001300898.1 Intron NP_001287827.1
XM_017007717.1 Intron XP_016863206.1
XM_017007718.1 Intron XP_016863207.1

View Full Product Details