Product Details
- SNP ID
-
rs142479343
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84978521 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCTACAGCTTTCATGGGGAGCAT[A/G]GCCATGACAACACCTTAATTTCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDFY3-AS2
PubMed Links
Gene Details
- Gene
- WDFY3-AS2
- Gene Name
- WDFY3 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details