Product Details

SNP ID
rs143969070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139293681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGAGGAAGTCCCAAACCCAT[C/T]AGATAAAAAACAAAATTTTAAATAT
Phenotype
MIM: 608000 MIM: 603844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAA15 PubMed Links

Gene Details

Gene
NAA15
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
There are no transcripts associated with this gene.

Gene
NDUFC1
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184986.1 Intron NP_001171915.1
NM_001184987.1 Intron NP_001171916.1
NM_001184988.1 Intron NP_001171917.1
NM_001184989.1 Intron NP_001171918.1
NM_001184990.1 Intron NP_001171919.1
NM_001184991.1 Intron NP_001171920.1
NM_002494.3 Intron NP_002485.1

View Full Product Details