Product Details

SNP ID
rs145837106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127881144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGAGCCTGGAAATATGGCGACC[A/T]GCATCGGGGAGAAGATCGAGGTGAA
Phenotype
MIM: 605031
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLK4 PubMed Links

Gene Details

Gene
PLK4
Gene Name
polo like kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190799.1 246 Missense Mutation AGC,TGC S4C NP_001177728.1
NM_001190801.1 246 Intron NP_001177730.1
NM_014264.4 246 Missense Mutation AGC,TGC S4C NP_055079.3
XM_005262701.2 246 Missense Mutation AGC,TGC S4C XP_005262758.1
XM_017007662.1 246 Intron XP_016863151.1
XM_017007663.1 246 Intron XP_016863152.1

View Full Product Details