Product Details

SNP ID
rs147543296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68229267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAAAAAACTTACCATTCATATA[A/T]AGTGTTCCCCAACCTGTAACTACAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11B PubMed Links

Gene Details

Gene
TMPRSS11B
Gene Name
transmembrane protease, serine 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182502.3 908 Silent Mutation CTA,CTT L312L NP_872308.2
XM_011531608.2 908 Silent Mutation CTA,CTT L312L XP_011529910.1
XM_011531609.1 908 UTR 3 XP_011529911.1

View Full Product Details