Product Details
- SNP ID
-
rs148072754
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112514726 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTGTCGGGAGGTGGGGGCTGCCA[A/G]TAGTCCATGTCTGACCCGGCCGGGC
- Phenotype
-
MIM: 606624
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEUROG2
PubMed Links
Gene Details
- Gene
- NEUROG2
- Gene Name
- neurogenin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024019.3 |
1077 |
Silent Mutation |
TAC,TAT |
Y250Y |
NP_076924.1 |
View Full Product Details