Product Details

SNP ID
rs138469332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81987610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATTGGAGAAAAAACATTCCAA[C/T]GTTATTGTGCAGAATTCATTAAACA
Phenotype
MIM: 610800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG10 PubMed Links

Gene Details

Gene
ATG10
Gene Name
autophagy related 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131028.1 338 Missense Mutation CGT,TGT R14C NP_001124500.1
NM_031482.4 338 Missense Mutation CGT,TGT R14C NP_113670.1
XM_005248610.4 338 Missense Mutation CGT,TGT R14C XP_005248667.1
XM_005248611.4 338 Intron XP_005248668.1
XM_005248612.3 338 Missense Mutation CGT,TGT R14C XP_005248669.1
XM_011543660.2 338 Intron XP_011541962.1
XM_011543661.2 338 Intron XP_011541963.1
XM_017009944.1 338 Missense Mutation CGT,TGT R14C XP_016865433.1

View Full Product Details