Product Details

SNP ID
rs139123635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58454673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGACATCAGCAGAGTTGTCAG[C/T]CTGAAAGTTGTAGATATCCTATCCT
Phenotype
MIM: 607023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLK2 PubMed Links

Gene Details

Gene
PLK2
Gene Name
polo like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252226.1 2148 Silent Mutation AGA,AGG R642R NP_001239155.1
NM_006622.3 2148 Silent Mutation AGA,AGG R656R NP_006613.2

View Full Product Details