Product Details

SNP ID
hCV171480435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140882346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGATCCTCGCAGCCTGGGAGAC[C/G]GGTAGTGGCCAGCTCCACTACTCCG
Phenotype
MIM: 606307 MIM: 606316 MIM: 606317 MIM: 606318 MIM: 606319 MIM: 606308 MIM: 606309 MIM: 606310 MIM: 606311 MIM: 606312 MIM: 606313 MIM: 606314 MIM: 606315
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
There are no transcripts associated with this gene.

Gene
PCDHA10
Gene Name
protocadherin alpha 10
There are no transcripts associated with this gene.

Gene
PCDHA11
Gene Name
protocadherin alpha 11
There are no transcripts associated with this gene.

Gene
PCDHA12
Gene Name
protocadherin alpha 12
There are no transcripts associated with this gene.

Gene
PCDHA13
Gene Name
protocadherin alpha 13
There are no transcripts associated with this gene.

Gene
PCDHA2
Gene Name
protocadherin alpha 2
There are no transcripts associated with this gene.

Gene
PCDHA3
Gene Name
protocadherin alpha 3
There are no transcripts associated with this gene.

Gene
PCDHA4
Gene Name
protocadherin alpha 4
There are no transcripts associated with this gene.

Gene
PCDHA5
Gene Name
protocadherin alpha 5
There are no transcripts associated with this gene.

Gene
PCDHA6
Gene Name
protocadherin alpha 6
There are no transcripts associated with this gene.

Gene
PCDHA7
Gene Name
protocadherin alpha 7
There are no transcripts associated with this gene.

Gene
PCDHA8
Gene Name
protocadherin alpha 8
There are no transcripts associated with this gene.

Gene
PCDHA9
Gene Name
protocadherin alpha 9
There are no transcripts associated with this gene.

View Full Product Details