Product Details

SNP ID
rs150746764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58454640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATTCCATTCGATTTTTTAATTC[C/T]GATGAACAGCCAGACATCAGCAGAG
Phenotype
MIM: 607023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLK2 PubMed Links

Gene Details

Gene
PLK2
Gene Name
polo like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252226.1 2181 Silent Mutation TCA,TCG S653S NP_001239155.1
NM_006622.3 2181 Silent Mutation TCA,TCG S667S NP_006613.2

View Full Product Details