Product Details

SNP ID
rs139455322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169217317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACTTCAAATTTGAATTGACCCT[G/T]GTTTATAATTTATTTAATGGGATTT
Phenotype
MIM: 188061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929523 PubMed Links

Gene Details

Gene
LOC101929523
Gene Name
uncharacterized LOC101929523
There are no transcripts associated with this gene.

Gene
THBS2
Gene Name
thrombospondin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003247.3 4344 UTR 3 NP_003238.2

View Full Product Details