Product Details
- SNP ID
-
rs141948771
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:85507988 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTACCTGCTTGTTGAGTACTGGTT[C/G]CTGTAAGCCATCAAACAGAAGTCTG
- Phenotype
-
MIM: 616105
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX14
PubMed Links
Gene Details
- Gene
- SNX14
- Gene Name
- sorting nexin 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001297614.2 |
2874 |
Missense Mutation |
CAA,GAA |
Q900E |
NP_001284543.1 |
NM_001304479.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q857E |
NP_001291408.1 |
NM_020468.5 |
2874 |
Missense Mutation |
CAA,GAA |
Q856E |
NP_065201.1 |
NM_153816.5 |
2874 |
Missense Mutation |
CAA,GAA |
Q909E |
NP_722523.1 |
XM_005248738.3 |
2874 |
Missense Mutation |
CAA,GAA |
Q908E |
XP_005248795.1 |
XM_005248740.3 |
2874 |
Missense Mutation |
CAA,GAA |
Q865E |
XP_005248797.1 |
XM_011535977.2 |
2874 |
Intron |
|
|
XP_011534279.1 |
XM_011535979.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q813E |
XP_011534281.1 |
XM_017011090.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q856E |
XP_016866579.1 |
XM_017011091.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q899E |
XP_016866580.1 |
XM_017011092.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q864E |
XP_016866581.1 |
XM_017011093.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q856E |
XP_016866582.1 |
XM_017011094.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q812E |
XP_016866583.1 |
XM_017011095.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q559E |
XP_016866584.1 |
XM_017011096.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q524E |
XP_016866585.1 |
XM_017011097.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q524E |
XP_016866586.1 |
XM_017011098.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q524E |
XP_016866587.1 |
XM_017011099.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q524E |
XP_016866588.1 |
XM_017011100.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q524E |
XP_016866589.1 |
XM_017011101.1 |
2874 |
Missense Mutation |
CAA,GAA |
Q515E |
XP_016866590.1 |
View Full Product Details