Product Details

SNP ID
rs142122809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131637410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAATCTACGTTGACTTTAGCAA[C/T]GGAACAACCTGTTAAGAAGAACACT
Phenotype
MIM: 602182 MIM: 605042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENPP3 PubMed Links

Gene Details

Gene
ENPP3
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005021.4 176 Missense Mutation ACG,ATG T9M NP_005012.2
XM_011535897.1 176 Intron XP_011534199.1
XM_017010932.1 176 Intron XP_016866421.1
Gene
MED23
Gene Name
mediator complex subunit 23
There are no transcripts associated with this gene.

View Full Product Details