Product Details

SNP ID
rs144658876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41229533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTCTTTAGCCCCAACCACGT[C/T]TCCTATGTGGACTCTTCCCTGGCTC
Phenotype
MIM: 614664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TREML4 PubMed Links

Gene Details

Gene
TREML4
Gene Name
triggering receptor expressed on myeloid cells like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198153.2 511 Missense Mutation TCT,TTT S136F NP_937796.1

View Full Product Details