Product Details

SNP ID
rs145560714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35260998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTAACATGTTTGCAAAGCAGTA[C/T]AGCAGAGTGCTTCATTTGGGGGCAT
Phenotype
MIM: 614708 MIM: 194549
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCUBE3 PubMed Links

Gene Details

Gene
SCUBE3
Gene Name
signal peptide, CUB domain and EGF like domain containing 3
There are no transcripts associated with this gene.

Gene
ZNF76
Gene Name
zinc finger protein 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292032.1 Intron NP_001278961.1
NM_003427.4 Intron NP_003418.2
XM_011514846.2 Intron XP_011513148.1
XM_011514847.2 Intron XP_011513149.1
XM_011514849.2 Intron XP_011513151.1
XM_011514850.2 Intron XP_011513152.1
XM_011514854.2 Intron XP_011513156.1
XM_011514855.2 Intron XP_011513157.1
XM_011514856.2 Intron XP_011513158.1
XM_017011248.1 Intron XP_016866737.1
XM_017011249.1 Intron XP_016866738.1
XM_017011250.1 Intron XP_016866739.1
XM_017011251.1 Intron XP_016866740.1
XM_017011252.1 Intron XP_016866741.1
XM_017011253.1 Intron XP_016866742.1
XM_017011254.1 Intron XP_016866743.1
XM_017011255.1 Intron XP_016866744.1
XM_017011256.1 Intron XP_016866745.1
XM_017011257.1 Intron XP_016866746.1

View Full Product Details