Product Details

SNP ID
rs146880588
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137493529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGTGGAGGGCGCCTTGAGTAG[A/C]GAGTGGGGAGGCCGGATGGTGCCGA
Phenotype
MIM: 609323
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
OLIG3 PubMed Links

Gene Details

Gene
OLIG3
Gene Name
oligodendrocyte transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175747.2 866 Silent Mutation TCG,TCT S214S NP_786923.1

View Full Product Details