Product Details

SNP ID
rs147403459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25726159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCTTTGGGCTTTATGGTGGTG[A/T]CTCTCAGTCTTCTTGGGCAGCAGCA
Phenotype
MIM: 613499 MIM: 609904 MIM: 182308
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AA PubMed Links

Gene Details

Gene
HIST1H2AA
Gene Name
histone cluster 1, H2aa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170745.3 404 Missense Mutation AGA,AGT R123S NP_734466.1
Gene
HIST1H2APS1
Gene Name
histone cluster 1, H2a, pseudogene 1
There are no transcripts associated with this gene.

Gene
HIST1H2BA
Gene Name
histone cluster 1, H2ba
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170610.2 404 Intron NP_733759.1
Gene
SLC17A1
Gene Name
solute carrier family 17 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005074.3 404 Intron NP_005065.2
XM_011514818.2 404 Intron XP_011513120.2
XM_011514819.2 404 Intron XP_011513121.2
XM_011514820.2 404 Intron XP_011513122.2
XM_011514821.2 404 Intron XP_011513123.1
XM_017011199.1 404 Intron XP_016866688.1
XM_017011200.1 404 Intron XP_016866689.1
XM_017011201.1 404 Intron XP_016866690.1
XM_017011202.1 404 Intron XP_016866691.1

View Full Product Details