Product Details

SNP ID
rs147952414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30743982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGGAAAGTGCTGAGGTCCAGA[A/G]CGTAGTCCGAGGGCTCCGAAGTCAG
Phenotype
MIM: 606998 MIM: 602996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLOT1 PubMed Links

Gene Details

Gene
FLOT1
Gene Name
flotillin 1
There are no transcripts associated with this gene.

Gene
HCG20
Gene Name
HLA complex group 20 (non-protein coding)
There are no transcripts associated with this gene.

Gene
IER3
Gene Name
immediate early response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003897.3 457 Missense Mutation GCT,GTT A142V NP_003888.2

View Full Product Details