Product Details

SNP ID
rs148116183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132617655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATAGCCCTCTATTCCATCTGCA[C/T]AGGCCTCTGAGAAGACCACCCCGAA
Phenotype
MIM: 604849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR2 PubMed Links

Gene Details

Gene
TAAR2
Gene Name
trace amine associated receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033080.1 439 Missense Mutation NP_001028252.1
NM_014626.3 439 Missense Mutation NP_055441.2
Gene
TAAR3
Gene Name
trace amine associated receptor 3 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details