Product Details
- SNP ID
-
rs150015158
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:41228929 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAGCCCAATGCTGGCTTCTTCAA[C/T]ATCACCATGATTCAGCTGACACAGA
- Phenotype
-
MIM: 614664
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TREML4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2101269] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TREML4
- Gene Name
- triggering receptor expressed on myeloid cells like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198153.2 |
383 |
Silent Mutation |
AAC,AAT |
N93N |
NP_937796.1 |
View Full Product Details