Product Details

SNP ID
rs138202050
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76400374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACTCGCTCAGCGCAGCTTCCTG[C/T]CCGCGGCACTCCACGTTGTCCAGCA
Phenotype
MIM: 607639 MIM: 182889
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SSC4D PubMed Links
Additional Information
For this assay, SNP(s) [rs4728712] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SSC4D
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080744.1 735 Silent Mutation GGA,GGG G129G NP_542782.1
XM_017011750.1 735 Intron XP_016867239.1
Gene
ZP3
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110354.1 735 Intron NP_001103824.1
NM_007155.5 735 Intron NP_009086.4
XM_017012586.1 735 Intron XP_016868075.1

View Full Product Details